Valganciclovir hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A247777-100MG
A247777-10MG
A247777-250MG
A247777-50MG
A247777-1G
A247777-25G
A247777-5G
A247777-5MG
76786-722EA
25
USD
76786-722
76786-724
76786-726
76786-728
77600-994
77668-018
77668-020
77668-022
Valganciclovir hydrochloride 98%
Valganciclovir hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₂₂N₆O₅·HCl |
MDL Number:
MFCD08460118 CAS Number: 175865-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...