4-Amino-1-methyl-1H-pyrazole hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-pyrazol-4-amine hydrochloride

A247043-100G A247043-10G A247043-1G A247043-25G A247043-5G
76786-712EA 241.89 USD
76786-712 76786-714 76786-716 76786-718 76786-720
4-Amino-1-methyl-1H-pyrazole hydrochloride 95%
4-Amino-1-methyl-1H-pyrazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClN₃
MW: 133.5794 g/mol
MDL Number: MFCD07626616
CAS Number: 127107-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR