Z-D-Dab-OH 98%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Amino-2-(((benzyloxy)carbonyl)amino)butanoic acid, Cbz-D-α-Dab-OH

A245524-1G A245524-250MG A245524-5G
76786-660EA 6.37 USD
76786-660 76786-662 76786-664 76786-666
Z-D-Dab-OH 98%
Z-D-Dab-OH

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₄
MDL Number: MFCD00237344
CAS Number: 70882-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR