4-(6-Chloro-2-trifluoromethylpyrimidin-4-yl)morpholine 95%

Supplier: Ambeed
Warning

A244500-100MG A244500-1G
76786-646EA 40.32 USD
76786-646 76786-648
4-(6-Chloro-2-trifluoromethylpyrimidin-4-yl)morpholine 95%
4-(6-Chloro-2-trifluoromethylpyrimidin-4-yl)morpholine

Precaución: for Research Use Only

Formula: C₉H₉ClF₃N₃O
MDL Number: MFCD09743689
CAS Number: 1189444-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR