tert-Butyl 3-(6-amino-3-methylpyridin-2-yl)benzoate 98%

Supplier: Ambeed
Warning

A244133-100MG A244133-250MG A244133-1G A244133-5G
76786-634EA 28.65 USD
76786-634 76786-636 77600-972 77600-974
tert-Butyl 3-(6-amino-3-methylpyridin-2-yl)benzoate 98%
tert-Butyl 3-(6-amino-3-methylpyridin-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₂O₂
MW: 284.35 g/mol
MDL Number: MFCD16659611
CAS Number: 1083057-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR