5-((tert-Butoxycarbonyl)amino)pentyl 4-methylbenzenesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: 5-(t-Boc-amino)-1-pentyl-p-toluenesulphonate

Stabilization: Stabilized with TBC

A244026-1G A244026-250MG A244026-25G A244026-5G
76786-630EA 38.2 USD
76786-630 76786-632 77667-996 77667-998
5-((tert-Butoxycarbonyl)amino)pentyl 4-methylbenzenesulfonate 97%
5-((tert-Butoxycarbonyl)amino)pentyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₇H₂₇NO₅S
MW: 357.46 g/mol
MDL Number: MFCD07776967
CAS Number: 118811-34-0
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR