3-(Aminomethyl)benzonitrile 97%

Supplier: Ambeed
Warning

A243632-1G A243632-250MG A243632-25G A243632-5G
76786-622EA 26 USD
76786-622 76786-624 76786-626 76786-628
3-(Aminomethyl)benzonitrile 97%
3-(Aminomethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MDL Number: MFCD06797832
CAS Number: 10406-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR