Zinc stearate ZnO 12.5-14%

Supplier: Ambeed
Warning

Synonyms: Stearic acid zinc salt, Zinc(II) stearate

A243623-25G A243623-1000G A243623-100G A243623-500G
76786-620EA 6.37 USD
76786-620 77667-984 77667-986 77667-988
Zinc stearate ZnO 12.5-14%
Zinc stearate

Precaución: for Research Use Only

Formula: C₃₆H₇₀O₄Zn
MW: 632.34 g/mol
MDL Number: MFCD00013031
CAS Number: 557-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR