Methyl-3-(4-bromophenyl)acrylate 98%

Supplier: Ambeed
Warning

A243398-100G A243398-10G A243398-1G A243398-5G A243398-25G
76786-610EA 318 USD
76786-610 76786-612 76786-614 76786-616 76786-618 77872-223
Methyl-3-(4-bromophenyl)acrylate 98%
Methyl-3-(4-bromophenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MDL Number: MFCD00157162
CAS Number: 3650-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR