4-Bromo-4'-propylbiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-4'-propyl-1,1'-biphenyl

A243284-100G A243284-25G A243284-5G
76786-594EA 108.22 USD
76786-594 76786-596 76786-598
4-Bromo-4'-propylbiphenyl 98%
4-Bromo-4'-propylbiphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br
MW: 275.1800 g/mol
MDL Number: MFCD00060105
CAS Number: 58743-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR