1-(4-Chloropyridin-2-yl)ethanone 97%

Supplier: Ambeed
Warning

A243174-100MG A243174-250MG A243174-5G A243174-1G
76786-588EA 25 USD
76786-588 76786-590 76786-592 77593-550
1-(4-Chloropyridin-2-yl)ethanone 97%
1-(4-Chloropyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MDL Number: MFCD10697662
CAS Number: 60159-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR