(R)-Piperidine-3-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A242764-10G A242764-250MG A242764-5G A242764-1G A242764-25G
76786-582EA 57 USD
76786-582 76786-584 76786-586 77667-982 77872-216
(R)-Piperidine-3-carboxylic acid hydrochloride 95%
(R)-Piperidine-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MDL Number: MFCD00211280
CAS Number: 885949-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR