Succinic acid 99%

Supplier: Ambeed
Warning

A242621-1000G A242621-100G A242621-25G A242621-500G
76786-572EA 19 USD
76786-572 76786-574 76786-576 76786-578 76786-580
Succinic acid 99%
Succinic acid

Precaución: for Research Use Only

Formula: HOOCCH₂CH₂COOH
MDL Number: MFCD00002789
CAS Number: 110-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR