Zinc p-toluenesulfonate hydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Zinc p-toluenesulphonate xhydrate
A242272-1000G
A242272-100G
A242272-25G
A242272-500G
76786-562EA
63
USD
76786-562
76786-564
76786-566
76786-568
Zinc p-toluenesulfonate hydrate 98%
Zinc p-toluenesulfonate hydrate
Precaución: for Research Use Only
Formula:
C₁₄H₁₆O₇S₂Zn MW: 425.775 g/mol |
MDL Number:
MFCD00064811 CAS Number: 123334-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...