trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans-4-[(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexanecarboxylic acid, Mal-AMCHC, trans-4-((2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexanecarboxylic acid
A241752-10G
A241752-250MG
A241752-25G
A241752-5G
A241752-100MG
76786-550EA
140.04
USD
76786-550
76786-552
76786-554
76786-556
77667-978
trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 97%
trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₄ MW: 237.26 g/mol |
MDL Number:
MFCD11519178 CAS Number: 69907-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...