Taurolidine 97%

Supplier: Ambeed
Warning

A241743-25MG A241743-5G A241743-5MG A241743-1G A241743-1MG A241743-25G A241743-100G
76786-544EA 8 USD
76786-544 76786-546 76786-548 77600-948 77600-950 77600-952 77872-209
Taurolidine 97%
Taurolidine

Precaución: for Research Use Only

Formula: C₇H₁₆N₄O₄S₂
MDL Number: MFCD00865076
CAS Number: 19388-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR