Tetraheptylammonium bromide 98%
Supplier: Ambeed
|
Warning
|
A241408-100G
A241408-1G
A241408-25G
A241408-5G
76786-528EA
200
USD
76786-528
76786-530
76786-532
76786-534
Tetraheptylammonium bromide 98%
Tetraheptylammonium bromide
Precaución: for Research Use Only
Formula:
C₂₈H₆₀BrN MW: 490.7 g/mol Melting Pt: 96 |
MDL Number:
MFCD00011861 CAS Number: 4368-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...