Tetraethylammonium hydrogen sulphate 99%

Supplier: Ambeed
Warning

Synonyms: Tetraethylammonium bisulphate, Tetraethylammonium hydrogensulphate

A241378-5G A241378-25G
76786-526EA 7 USD
76786-526 77600-946
Tetraethylammonium hydrogen sulphate 99%
Tetraethylammonium hydrogen sulphate

Precaución: for Research Use Only

Formula: C₈H₂₁NO₄S
MW: 227.33
MDL Number: MFCD00036150
CAS Number: 16873-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR