Methyl 2-bromo-5-cyanobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-(methoxycarbonyl)benzonitrile

A240414-100MG A240414-1G A240414-250MG A240414-5G A240414-25G
76786-510EA 18.04 USD
76786-510 76786-512 76786-514 76786-516 77667-974
Methyl 2-bromo-5-cyanobenzoate 97%
Methyl 2-bromo-5-cyanobenzoate

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.053 g/mol
MDL Number: MFCD11035757
CAS Number: 1031927-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR