4-Bromo-2,5-dichloroaniline 97%

Supplier: Ambeed
Warning

A239512-10G A239512-1G A239512-25G A239512-5G
76786-496EA 22 USD
76786-496 76786-498 76786-500 76786-502
4-Bromo-2,5-dichloroaniline 97%
4-Bromo-2,5-dichloroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrCl₂N
MDL Number: MFCD08444535
CAS Number: 1940-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR