7-(Diethylamino)coumarin-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7ACC1, 7-(Diethylamino)-2-oxo-2H-chromene-3-carboxylic acid, DEAC, acid
A239506-100MG
A239506-1G
A239506-250MG
A239506-25G
A239506-25MG
A239506-5G
A239506-100G
76786-484EA
7.43
USD
76786-484
76786-486
76786-488
76786-490
76786-492
76786-494
77600-928
7-(Diethylamino)coumarin-3-carboxylic acid 98%
7-(Diethylamino)coumarin-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₅NO₄ MW: 261.28 g/mol |
MDL Number:
MFCD00068721 CAS Number: 50995-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...