trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol 98%

Supplier: Ambeed
Warning

Synonyms: (trans,trans)-4'-Pentyl-[1,1'-bi(cyclohexan)]-4-ol, trans-4-(trans-4-Amylcyclohexyl)cyclohexanol, trans,trans-4'-Amyl-[1,1'-bi(cyclohexane)]-4-ol, trans,trans-4'-Pentyl-[1,1'-bi(cyclohexane)]-4-ol

A239301-100G A239301-10G A239301-1G A239301-25G
76786-470EA 413.76 USD
76786-470 76786-472 76786-474 76786-476
trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol 98%
trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₇H₃₂O
MW: 252.44 g/mol
MDL Number: MFCD09752070
CAS Number: 82575-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR