4-Amino-3-chloropyridin-2-ol 97%

Supplier: Ambeed
Warning

A237829-100MG A237829-1G A237829-250MG
76786-432EA 112.46 USD
76786-432 76786-434 76786-436
4-Amino-3-chloropyridin-2-ol 97%
4-Amino-3-chloropyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.55 g/mol
MDL Number: MFCD09763693
CAS Number: 55290-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR