trans-2-Decenal 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
trans-2-Decenal, (E)-2-Decenal, (E)-Dec-2-enal
A237278-5G
A237278-100G
76786-430EA
15.92
USD
76786-430
77667-950
trans-2-Decenal 98%
trans-2-Decenal
Precaución: for Research Use Only
Formula:
C₁₀H₁₈O MW: 154.25 g/mol |
MDL Number:
MFCD00014679 CAS Number: 3913-81-3 UN: 3082 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...