(1R,2R)-rel-1,2-Diphenylethane-1,2-diamine 98%

Supplier: Ambeed
Warning

A236727-10G A236727-1G A236727-25G
76786-420EA 127.31 USD
76786-420 76786-422 76786-424
(1R,2R)-rel-1,2-Diphenylethane-1,2-diamine 98%
(1R,2R)-rel-1,2-Diphenylethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂
MW: 212.29 g/mol
MDL Number: MFCD00082751
CAS Number: 16635-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR