Methyl-3-amino-6-iodopyrazine-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-6-iodopyrazine-2-carboxylate

A234499-10G A234499-1G A234499-250MG A234499-5G
76786-376EA 385.11 USD
76786-376 76786-378 76786-380 76786-382
Methyl-3-amino-6-iodopyrazine-2-carboxylate 95%
Methyl-3-amino-6-iodopyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆IN₃O₂
MW: 279.04 g/mol
MDL Number: MFCD09909640
CAS Number: 1458-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR