(S)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A234482-1G A234482-250MG A234482-25G A234482-5G
76786-368EA 21.22 USD
76786-368 76786-370 76786-372 76786-374
(S)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid 97%
(S)-1-((Benzyloxy)carbonyl)piperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₄
MW: 263.29 g/mol
MDL Number: MFCD04114968
CAS Number: 88466-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR