(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1R,2S)-1-Boc-2-vinylcyclopropanecarboxylic acid, (1R,2S)-1-((tert-Butoxycarbonyl)amino)-2-vinylcyclopropanecarboxylic acid, (1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-ethenylcyclopropanecarboxylic acid

A234091-1G A234091-250MG A234091-5G
76786-362EA 33.95 USD
76786-362 76786-364 76786-366
(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic acid 95%
(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD11042412
CAS Number: 159622-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR