5-Bromo-2-(trifluoromethoxy)benzoic acid 98%

Supplier: Ambeed
Warning

A233243-1G A233243-25G A233243-5G
76786-340EA 41.38 USD
76786-340 76786-342 76786-344
5-Bromo-2-(trifluoromethoxy)benzoic acid 98%
5-Bromo-2-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₃
MW: 285.015 g/mol
MDL Number: MFCD06203214
CAS Number: 403646-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR