Ethyl 4-nitro-1H-pyrazole-3-carboxylate 95%

Supplier: Ambeed
Warning

A232918-1G A232918-250MG A232918-25G A232918-5G
76786-314EA 37.14 USD
76786-314 76786-316 76786-318 76786-320
Ethyl 4-nitro-1H-pyrazole-3-carboxylate 95%
Ethyl 4-nitro-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₄
MW: 185.14 g/mol
MDL Number: MFCD00195040
CAS Number: 55864-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR