2,6-Naphthyridin-1(2H)-one 97%

Supplier: Ambeed
Warning

A232535-100MG A232535-1G A232535-250MG A232535-5G
76786-302EA 43 USD
76786-302 76786-304 76786-306 77872-178
2,6-Naphthyridin-1(2H)-one 97%
2,6-Naphthyridin-1(2H)-one

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MDL Number: MFCD06762736
CAS Number: 80935-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR