Trimethoprim lactate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(3,4,5-Trimethoxybenzyl)pyrimidine-2,4-diamine 2-hydroxypropanoate
A232194-100G
A232194-10G
A232194-25G
76786-296EA
82.76
USD
76786-296
76786-298
76786-300
Trimethoprim lactate 97%
Trimethoprim lactate
Precaución: for Research Use Only
Formula:
C₁₇H₂₄N₄O₆ MW: 380.401 g/mol |
MDL Number:
MFCD00171722 CAS Number: 23256-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...