6-Iodoisobenzofuran-1(3H)-one 95%

Supplier: Ambeed
Warning

A231995-100MG A231995-1G A231995-250MG
76786-286EA 28 USD
76786-286 76786-288 76786-290
6-Iodoisobenzofuran-1(3H)-one 95%
6-Iodoisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅IO₂
MDL Number: MFCD06739592
CAS Number: 53910-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR