2-Amino-5-bromo-4-dimethoxymethylpyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-(dimethoxymethyl)pyrimidin-2-amine

A231429-100MG A231429-1G A231429-250MG A231429-5G
76786-274EA 25.47 USD
76786-274 76786-276 76786-278 76786-280
2-Amino-5-bromo-4-dimethoxymethylpyrimidine 97%
2-Amino-5-bromo-4-dimethoxymethylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₁₀BrN₃O₂
MW: 248.09 g/mol
MDL Number: MFCD08460359
CAS Number: 914347-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR