(S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol 95%

Supplier: Ambeed
Warning

A231146-100G A231146-10G A231146-1G A231146-250MG A231146-25G A231146-5G
76786-256EA 214.31 USD
76786-256 76786-258 76786-260 76786-262 76786-264 76786-266
(S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol 95%
(S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇Cl₂FO
MW: 209.04 g/mol
MDL Number: MFCD09863793
CAS Number: 877397-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR