Tenoxicam 98%

Supplier: Ambeed
Warning

A230274-100G A230274-1G A230274-250MG A230274-25G A230274-5G
76786-234EA 228 USD
76786-234 76786-236 76786-238 76786-240 76786-242
Tenoxicam 98%
Tenoxicam

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃O₄S₂
MDL Number: MFCD00083502
CAS Number: 59804-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR