2,4,6-Trichlorobenzyl alcohol 97%

Supplier: Ambeed
Warning

A230085-1G A230085-250MG A230085-5G
76786-224EA 20 USD
76786-224 76786-226 76786-228
2,4,6-Trichlorobenzyl alcohol 97%
2,4,6-Trichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅Cl₃O
MDL Number: MFCD03093891
CAS Number: 217479-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR