4-[2-(Trimethylsilyl)ethoxycarbonyloxy]nitrobenzene 97%

Supplier: Ambeed
Warning

A229619-1G A229619-250MG A229619-5G A229619-10G A229619-25G
76786-212EA 25 USD
76786-212 76786-214 76786-216 77593-482 77872-166
4-[2-(Trimethylsilyl)ethoxycarbonyloxy]nitrobenzene 97%
4-[2-(Trimethylsilyl)ethoxycarbonyloxy]nitrobenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₅Si
MDL Number: MFCD00042930
CAS Number: 80149-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR