2-Bromo-6-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A229613-100MG A229613-1G A229613-250MG A229613-5G
76786-204EA 35.01 USD
76786-204 76786-206 76786-208 76786-210
2-Bromo-6-methoxypyrimidine 98%
2-Bromo-6-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MW: 189.0100 g/mol
MDL Number: MFCD09702031
CAS Number: 944709-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR