5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine 98%

Supplier: Ambeed
Warning

A229224-100MG A229224-1G A229224-250MG A229224-25G A229224-5G
76786-190EA 16.98 USD
76786-190 76786-192 76786-194 76786-196 76786-198
5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine 98%
5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrIN₃
MW: 323.9166 g/mol
MDL Number: MFCD09878697
CAS Number: 875781-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR