7-Bromo-2-chloroquinazoline 95%

Supplier: Ambeed
Warning

A228637-100MG A228637-1G A228637-250MG
76786-184EA 24.41 USD
76786-184 76786-186 76786-188
7-Bromo-2-chloroquinazoline 95%
7-Bromo-2-chloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MW: 243.4900 g/mol
MDL Number: MFCD11040175
CAS Number: 953039-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR