5-Ethylpiperidine-2,4-dione 95%

Supplier: Ambeed
Warning

A227943-100MG A227943-1G A227943-250MG
76786-172EA 51.99 USD
76786-172 76786-174 76786-176
5-Ethylpiperidine-2,4-dione 95%
5-Ethylpiperidine-2,4-dione

Precaución: for Research Use Only

Formula: C7H11NO2
MW: 141.1700 g/mol
MDL Number: MFCD09999166
CAS Number: 73290-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR