Triethylvinylsilane 98+% stabilized

Supplier: Ambeed
Danger

Synonyms: Vinyltriethylsilane

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A227693-1G A227693-5G
76786-164EA 16.98 USD
76786-164 76786-166
Triethylvinylsilane 98+% stabilized
Triethylvinylsilane

Precaución: for Research Use Only

Formula: C₈H₁₈Si
MW: 142.32 g/mol
MDL Number: MFCD00041345
CAS Number: 1112-54-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR