6-Bromo-1,8-naphthyridin-2-amine 98%

Supplier: Ambeed
Warning

A227352-100MG A227352-1G A227352-250MG A227352-5G
76786-154EA 29 USD
76786-154 76786-156 76786-158 76786-160
6-Bromo-1,8-naphthyridin-2-amine 98%
6-Bromo-1,8-naphthyridin-2-amine

Precaución: for Research Use Only

Formula: C₈H₆BrN₃
MDL Number: MFCD09909648
CAS Number: 64874-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR