2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1,3,7-triazaspiro[4.5]dec-2-en-4-one hydrochloride, 2-Methyl-4-oxo-1,3,7-triazaspiro[4.5]dec-1-ene hydrochloride

A227155-250MG A227155-100MG
76786-144EA 56.23 USD
76786-144 77667-888
2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride 97%
2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₃N₃O·ClH
MW: 203.669 g/mol
MDL Number: MFCD11869758
CAS Number: 1214028-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR