Isochroman-4-one 96%

Supplier: Ambeed
Warning

A224267-100MG A224267-1G A224267-250MG A224267-5G
76786-098EA 39 USD
76786-098 76786-100 76786-102 76786-104
Isochroman-4-one 96%
Isochroman-4-one

Precaución: for Research Use Only

Formula: C₉H₈O₂
MDL Number: MFCD09744058
CAS Number: 20924-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR