BEZ235 98+%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-2-(4-(3-methyl-2-oxo-8-(quinolin-3-yl)-2,3-dihydro-1H-imidazo[4,5-c]quinolin-1-yl)phenyl)propanenitrile, NVP-BEZ235, Dactolisib

A224243-100MG A224243-1MG A224243-50MG A224243-10MG
76786-086EA 114.58 USD
76786-086 76786-088 76786-090 77600-802
BEZ235 98+%
BEZ235

Precaución: for Research Use Only

Formula: C₃₀H₂₃N₅O
MW: 469.6 g/mol
MDL Number: MFCD10565944
CAS Number: 915019-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR