4-Bromo-2-(trifluoromethoxy)benzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A224072-100G
A224072-10G
A224072-1G
A224072-250MG
A224072-25G
A224072-5G
76786-074EA
530.45
USD
76786-074
76786-076
76786-078
76786-080
76786-082
76786-084
4-Bromo-2-(trifluoromethoxy)benzaldehyde 97%
4-Bromo-2-(trifluoromethoxy)benzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₄BrF₃O₂ MW: 269.0200 g/mol |
MDL Number:
MFCD12406886 CAS Number: 220996-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...