Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 95%
Supplier: Ambeed
|
Warning
|
A223874-1G
A223874-250MG
A223874-25G
A223874-5G
A223874-100G
76786-070EA
8
USD
76786-070
76786-072
77600-796
77600-798
77667-856
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 95%
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Precaución: for Research Use Only
Formula:
C₉H₁₂N₂Na₃O₁₅P₃ |
MDL Number:
MFCD00044310 CAS Number: 19817-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...