5-Iodopyrazin-2-amine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-iodopyrazine
A222855-100G
A222855-10G
A222855-1G
A222855-25G
A222855-5G
76786-048EA
654.58
USD
76786-048
76786-050
76786-052
76786-054
76786-056
5-Iodopyrazin-2-amine 95%
5-Iodopyrazin-2-amine
Precaución: for Research Use Only
Formula:
C4H4IN3 MW: 221.0000 g/mol |
MDL Number:
MFCD09909691 CAS Number: 886860-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...